Bottles, wide neck, amber, with GLS 80® thread and screw cap, DURAN®
供应商: DWK LIFE SCIENCES
1160146 1160151 1160150 1160149 1160148 1160147 218666959 218663653
SCOT1160146EA 0 CNY
SCOT1160146 SCOT1160151 SCOT1160150 SCOT1160149 SCOT1160148 SCOT1160147 SCOT218666959 SCOT218663653
Bottles, wide neck, amber, with GLS 80® thread and screw cap, DURAN®
玻璃瓶
DURAN®, borosilicate glass 3,3, amber, with blue PP screw cap.
  • Very good chemical resistance, high temperature resistance
  • Extra wide opening allows easy cleaning
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
  • Amber glass meets USP 671 and Eur. Ph. 3.2.1. for protection against UV light
Amber to protect light-sensitive materials. UV protection up to approx. 500 nm light wavelength. Only the outer surface of the bottles is tinted ensuring that the DURAN® properties on the inside remain unchanged. Round, with graduations and specially developed GLS 80® thread. Opening and closing the bottle is easy with quick closure mechanism. The 80 mm wide outer diameter of the bottle neck enables bottles to be filled conveniently and safely with powders, pastes, granulates and liquids. The large opening can accommodate even large volume funnels, and gives easy access to scoops or spatulas.
Med tryckt spårningskod (batchidentifiering).