Cell culture roller bottles, Corning®
供应商: CORNING SCIENCE PRODUCTS
430195 430699 430852 431135 431198 431191 430849 3907 431134 431344 431321 431329 430853 430851 431303
CORN430195EA 0 CNY
CORN430195 CORN430699 CORN430852 CORN431135 CORN431198 CORN431191 CORN430849 CORN3907 CORN431134 CORN431344 CORN431321 CORN431329 CORN430853 CORN430851 CORN431303
Cell culture roller bottles, Corning®
滚瓶
Virgin PS, sterile.
  • One piece seamless construction
  • All bottles have printed lot numbers to aid product traceability
  • Sterilised by gamma radiation
  • Certified non-pyrogenic
  • Ribbed design provides twice the surface area with the same exterior dimensions
Roller bottles available with smooth or ribbed surface design for expanded culture area. All bottles are treated for optimal cell attachment. The Corning® CellBIND® surface enhances cell attachment under difficult conditions, such as reduced-serum or serum-free medium, resulting in higher cell yields. All bottles are graduated, unless otherwise stated.
Note: Corning recommends 0,2 to 0,3 ml of medium per cm² of growth area, and setting roller rack speeds to provide 0,5 to 1,0 min⁻¹.
Caution: The product(s) on this page are outside the scope of the EU Directive 2019/904