VWR® Performance Manual Defrost Laboratory Freezers
供应商: VWR BRAND PRIVATE LABEL USA
10819-362 10819-366 10819-364 10819-368
VWRU10819-362EA 229489 CNY
VWRU10819-362 VWRU10819-366 VWRU10819-364 VWRU10819-368
VWR® Performance Manual Defrost Laboratory Freezers
冷柜 实验室通用冰箱
Performance Series manual defrost laboratory freezers feature a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies.
  • Sample-simulated temperature display
  • Alarm mute with ring back
  • Caster platform adds four inches to height
  • Inner 'fast-freeze compartments' with doors for temperature maintenance and recovery
A manual defrost design prevents product damaging temperature spikes from defrost cycles. Defrosting of the unit must be performed periodically to prevent severe ice build-up. Standard features package includes keyed door locks, caster platforms, sample-simulated probe, probe access port. A USB port is provided for easy transfer as well as software updates. It features alarm battery backup, remote contacts, alarm validation, data acquisition and 24/7 data logging.
Certifications: UL/C-UL listed.
Delivery information: Freezers are supplied with three-year parts and labour warranty, five years on compressor parts.