VWR®Signature™, Forced air safety ovens
供应商: VWR BRAND PRIVATE LABEL USA
52201-214 52201-216
VWRU52201-214EA 142967 CNY
VWRU52201-214 VWRU52201-216
VWR®Signature™, Forced air safety ovens
干燥箱 风扇辅助和自然对流炉
These microprocessor-controlled forced air safety ovens have heavy gauge steel exteriors and continuous welded seams and corners for long-term durability. The powder coat enamel exterior provides a high resistance to moisture, oil and chemicals.
  • Over-temperature safety limit, manual restart
  • Rear blow-out panel
The panel is designed to release in the event of an explosion, directing the force away from the user. A resettable circuit breaker prevents accidental setting changes in case of electrical malfunction. An over-temperature safety limit requiresMore Product Information
现在订购
规格
型号 1330FMS 1350FMS
Environmentally Preferable
Temperature fluctuation (time) ±1,9 @ 170 °C ±1,9 @ 170 °C
温度范围 40…240 °C 40…240 °C
容量 45 L 115 L
对流类型 强制对流 强制对流
Shelves supplied (max.) 2 2
Int. W×D×H 330×356×375 mm 457×483×419 mm
Ext. W×D×H 546×622×838 mm 700×686×883 mm
功耗 1750 W 2000 W