VWR® Performance, Glass door laboratory refrigerators with natural refrigerants
供应商: VWR BRAND PRIVATE LABEL USA
10820-344 10819-530
VWRU10820-344EA 364275 CNY
VWRU10820-344 VWRU10819-530
VWR® Performance, Glass door laboratory refrigerators with natural refrigerants
冰箱
These performance series glass door laboratory refrigerators are purpose-built for scientific and medical applications requiring superior temperature management and state of the art product security and data logging.
  • Adjustable operating temperature range: 1 to 10 °C
  • Sample simulated digital temperature display
  • Password-protected control panel
  • Audible and visual high/low temperature alarms
It features a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies. A forced air directional refrigeration system provides superior temperature uniformity and recovery after door openings. An automatic cycle-defrost design prevents product damaging temperature spikes. Additional safety features include power outage alarm, sensor error alarm, and door ajar alarm.
Remote alarm dry contacts and an access port are provided to simplify connections to existing facility monitoring systems.
Units are supplied with a three-years parts and labour warranty, with an additional five-years warranty on compressor parts.
Certifications: UL/C-UL Listed.