VWR® Talon®, Support stand, U-shaped base
供应商: VWR BRAND PRIVATE LABEL USA
14010-004 14010-002
VWRU14010-004EA 15128.85 CNY
VWRU14010-004 VWRU14010-002
VWR® Talon®, Support stand, U-shaped base
支撑架
This support stand with sturdy U-shaped base is entirely made of stainless steel (Type 303) and can accomodate cylindrical vessels up to 457 mm (18") in diameter.
  • Fitted with oversized knobs for secure positioning
  • Threaded stainless steel support rod is 16 mm (5/8") in diameter and screwed into the support stand base
Delivery information: Supplied with support rod.