VWR®, Crimp Top Vials, 11 mm
供应商: VWR BRAND PRIVATE LABEL USA
66010-539 46610-742
VWRU66010-539EA 573 CNY
VWRU66010-539 VWRU46610-742
VWR®, Crimp Top Vials, 11 mm
小瓶
Wide opening vials manufactured from 33 expansion borosilicate glass.
  • Easier access
  • Reduced needle damage
  • Amber vials for light-sensitive samples
  • 40% larger neck opening than standard vials