VWR®, Vacuum Ovens
供应商: VWR BRAND PRIVATE LABEL USA
89508-428 89508-426
VWRU89508-428EA 255684 CNY
VWRU89508-428 VWRU89508-426
VWR®, Vacuum Ovens
干燥箱 真空炉
VWR® vacuum ovens with powder-coated heavy-gauge steel exterior for durability, easy to clean Type 304 stainless-steel interior chamber, and glass wool insulation to prevent heat loss.
  • Heating system optimises chamber space for drying, curing, vacuum embedding and plating applications
  • Polycarbonate safety shield protects door glass
  • Built-in over-temperature protection
  • Corrosion-resistant stainless-steel tubing for long-term performance
  • With two removable stacking aluminum shelves
  • Silicone door gasket and positive latch door maintain seal at all vacuum levels
The 12,5 l unit feature top-mounted independent evacuation and venting vacuum fittings. The 19,8 l and 65,1 l units feature a front-mounted three-way valve for evacuation, venting and purging of inert gases (e.g. nitrogen or argon) with vacuum fittings on the front of the unit.
Delivery information: Available with two year parts and labor warranty.