VWR®, Media Bottles, Square, PETG, Sterile
供应商: VWR BRAND PRIVATE LABEL USA
89132-060 89132-062 89132-056 89132-058 89132-064 89132-066
VWRU89132-060EA 2645.86 CNY
VWRU89132-060 VWRU89132-062 VWRU89132-056 VWRU89132-058 VWRU89132-064 VWRU89132-066
VWR®, Media Bottles, Square, PETG, Sterile
培养基瓶
These square/octagonal PETG ergonomic media bottles are ideal for storage of media, laboratory reagents and biological buffers.
  • Graduation markings are moulded in for reference filling
  • Bottles are gamma sterilised (SAL 10⁻⁶), with blue PP caps attached
Packaging: Each case is specifically packaged for sterile production with multi-barrier elastomer packaging.