VWR®Standard, Solid door laboratory refrigerators
供应商: VWR BRAND PRIVATE LABEL USA
10820-402 10820-400
VWRU10820-402EA 0 CNY
VWRU10820-402 VWRU10820-400
VWR®Standard, Solid door laboratory refrigerators
冰箱
The standard solid door laboratory refrigerators are purpose-built for scientific and medical applications.
  • Adjustable operating temperature range: 1 to 10 °C
  • Sample simulated digital temperature display
  • Audible and visual high/low temperature alarms
It features a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies. A forced air directional refrigeration system provides superior temperature uniformity and recovery after door openings. An automatic cycle-defrost design prevents product damaging temperature spikes.
Remote alarm dry contacts and an access port are provided to simplify connections to existing facility monitoring systems.
Units are supplied with a two-year parts and labour warranty, with an additional three-year warranty on compressor parts.
Certifications: UL/C-UL Listed.