Ubbelohde viscometers, calibrated
供应商: TECHNICAL GLASS
UBB-1B UBB-2 UBB-3C UBB-3 UBB-1C
TEGPUBB-1BEA 0 CNY
TEGPUBB-1B TEGPUBB-2 TEGPUBB-3C TEGPUBB-3 TEGPUBB-1C
Ubbelohde viscometers, calibrated
粘度计 毛细管粘度计
These Ubbelohde viscometers are used to measure kinematic viscosity of transparent liquids.
  • Standard in petroleum industry
  • Well suited for use at very high and low temperatures
  • Viscometer constant does not vary with bath temperature
They suspend through a 510 mm hole of a constant temperature bath by means of a round holder and require a minimum sample of 11 ml and liquid bath depth of 241 mm.