VWR®, Bottle top vacuum filtration systems
供应商: VWR BRAND PRIVATE LABEL USA
97066-210 97066-204 97066-202
VWRU97066-210EA 7136 CNY
VWRU97066-210 VWRU97066-204 VWRU97066-202
VWR®, Bottle top vacuum filtration systems
过滤器 瓶口过滤器
These vacuum filtration systems are designed for the preparation of buffers, tissue culture media, microbiological media and other biological fluids. They feature large diameter Pall High Performance PES membranes for guaranteed performance and maximum throughput.
  • Quick-Connect™ pedestal base or cradle ring allows for hands-free filtration. Weighted for extra stability
  • Quick-Connect™ pedestal base ensures that the unit is always correctly placed
  • Forward-facing, large, raised graduation marks allow for easy visualisation of volumes
  • Vacuum on/off switch located at the front
  • Vacuum hose adapter to use with conventional filtration set-ups
The full assembly includes a filtration unit and a receiver bottle with an ergonomic shape, moulded finger grips and a patented bottle cap. All items are made from FDA grade, Class VI materials and are gamma sterilised VD Max 10⁻⁶ in a 100000 cleanroom in ISO 13485 standard facility.
订购须知 Vacuum filtration pedestal and vacuum filtration cradle ring are to be ordered separately.
Delivery information: Complete system includes filtration funnel (PS), media bottle (PS) and hose adapter.