二乙醚 96.5-100% stabilised USP, Macron Fine Chemicals™
供应商: Avantor Performance Materials
Danger
Stabilization: Stabilised with BHT (2,6-Di-tert-butyl-p-cresol, 2,6-Di-tert-butyl-4-methylphenol, Ionol) and ethanol
MACR0812-16EA 0 CNY
MACR0812-16 MACR0812-25
二乙醚 96.5-100% stabilised USP, Macron Fine Chemicals™
二乙醚
Contains BHT and ethanol as preservatives.
分子式: (CH₃CH₂)₂O
沸点: 35 °C (1013 hPa)
溶点: –123 °C
密度: 0,7135 g/cm³ (20 °C)
闪点: –40 °C
存储条件: Ambient
MDL编号: MFCD00011646
CAS号: 60-29-7
EINECS: 200-467-2
UN号: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM
现在订购
产品技术规格
Meets U.S.P Requirements 0.713 - 0.716
GMP Manufactured Product Passes Test
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical ≤0.2%
USP - Specific Gravity at 25°/25°C 0.713 - 0.716
USP - Acidity Passes Test
USP - Aldehydes Passes Test
USP - Low-Boiling Hydrocarbons ≤0.2 %
USP - Nonvolatile Residue ≤0.003 %
USP - Peroxide (as H₂O₂) ≤0.3 ppm
USP - Preservative (C₂H₅OH), For Information Only
USP - Preservative (H₂O), (w/w) ≤0.5 %
USP - Preservative (BHT) ≥7 ppm
USP - Water (H₂O)(by Karl Fischer titrn) ≤0.5 %
Only the Class 3 solvent Ethanol is likely to be present as a preservative. The actualconcentration is reported.