Sulphate TNTplus® kit
供应商: Hach
TNT865
TNT864
HACHTNT865EA
0
CNY
HACHTNT865
HACHTNT864
Sulphate TNTplus® kit
环境试验箱
This kit is used for the the determination of sulphate by the turbidimetric method using TNTplus vials.
- Reduce errors with bar coded vials and RFID
- Innovative vials with DosiCaps
- No reagent blank necessary