Measuring cylinders, graduated
供应商: HACH
50838 50849
HACH50838EA 0 CNY
HACH50838 HACH50849
Measuring cylinders, graduated
量筒 Measuring Cylinders
Graduated cylinders with white markings, Class B.
  • Made of glass
  • Calibrated 'To Contain'
  • With single metric scale in durable white markings