Cell proliferation assays, CellTiter 96® AQueous One Solution
供应商: Promega
G3580
G3581
PROMG3580EA
0
CNY
PROMG3580
PROMG3581
Cell proliferation assays, CellTiter 96® AQueous One Solution
检测产品
细胞化验
The CellTiter 96® AQueous One Solution Cell Proliferation Assay is a colorimetric method for determining the number of viable cells in proliferation, cytotoxicity or chemosensitivity assays.
- Simplify colorimetric viability assays
- Use as a single solution, filter sterilised and ready to add to assay plates
- No washing or cell harvesting and no solubilisation steps
- Plates can be read and returned to incubator for further colour development
- No volatile organic solvent is required to solubilise the formazan product
- No scintillation cocktail or radioactive waste disposal
The CellTiter 96® AQueous One Solution Cell Proliferation Assay is applicable for cell proliferation, cell viability, cytotoxicity, chemosensitivity, cell attachment determination and chemotaxis.