Stirring shafts, polished, CHEM-STIR™
供应商: CHEMGLASS
CG-2078-05 CG-2078-07
CGLACG-2078-05EA 0 CNY
CGLACG-2078-05 CGLACG-2078-07
Stirring shafts, polished, CHEM-STIR™
搅拌器 Stirring Shafts
Stirring shafts with a ring at the lower end to enable the blade to be attached by means of a PTFE nut and bolt . The polished shaft will not cause abrasion to the sealing surfaces of the PTFE bearing.
Delivery information: Shaft is supplied as a complete set with a PTFE nut and bolt. Stirrer blades must be ordered separately.