Pressure vessels
供应商: CHEMGLASS
CG-1880-05 CG-1880-10
CGLACG-1880-05EA 0 CNY
CGLACG-1880-05 CGLACG-1880-10
Pressure vessels
反应容器 圆柱形反应容器
These heavy wall pressure vessels have an internal thread for use with a 15 mm PTFE bushing as a pressure seal. This unique design allows hand tightening and loosing for easy access into the vessel.
  • Very good chemical resistance
  • Very good temperature resistance
The O-ring is positioned on the bushing above the thread for improved sealing. The vessels are useful for pressure reactions and applications such as polymerization and analysis of halogens by thermal decomposition in the presence of lime.
Delivery information: Complete set is supplied withMore Product Information