Standard polystyrene microplates, 1536-well, Corning®
供应商: CORNING SCIENCE PRODUCTS
3936 3937 3891 3893 3724 3724BC 3725 3725BC 3726 3727 3727BC 3728 3728BC 3729 3729BC 3891BC 3893BC 3895 3731BC 3838BC 3726BC
CORN3936EA 0 CNY
CORN3936 CORN3937 CORN3891 CORN3893 CORN3724 CORN3724BC CORN3725 CORN3725BC CORN3726 CORN3727 CORN3727BC CORN3728 CORN3728BC CORN3729 CORN3729BC CORN3891BC CORN3893BC CORN3895 CORN3731BC CORN3838BC CORN3726BC
Standard polystyrene microplates, 1536-well, Corning®
微孔板 高温超导微孔板
These PS microplates conform to standard microplate footprint and dimensions, and are available in solid black or white, with round or flat bottoms, or in black clear bottom formats.
  • Total well volume of 10 μl for round well plates and 12,8 μl for flat bottom plates
  • Recommended working volume of up to 8 μl
  • Round well bottoms for reduced air entrapment and improved CVs and Z factor
  • Raised well bottoms for higher sensitivity
  • Lids available separately
* NBS™ = Non binding surface