磷酸氢氨钠 ≥99.0%, Reagent Grade
供应商: VWR Chemicals
Synonyms:
磷酸氢铵钠, 磷酸氢钠铵, 磷酸钠铵
VWRC0770-P12KGEA
13764
CNY
VWRC0770-P12KG
VWRC0770-P25KG
磷酸氢氨钠 ≥99.0%, Reagent Grade
磷酸氢氨钠
分子式:
NaNH₄HPO₄·4H₂O 分子量: 209.07 g/mol 存储条件: Ambient |
MDL编号:
MFCD00066970 CAS号: 7783-13-3 |
产品技术规格
Arsenic | ≤0.0001% |
Assay | ≥99.0% |
Chloride | ≤0.0005% |
Heavy Metals | ≤0.0005% |
Insolubles | ≤0.01% |
Iron | ≤0.0005% |
Nitrate | ≤0.003% |
pH (5%, Water) at 25 °C | 7.5 – 8.5 |
Sulphate | ≤0.02% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...