酒石酸氢钠 98+% for analysis

供应商: Thermo Fisher Scientific

Synonyms: 酒石酸, L(+)-酒石酸, 重酒石酸钠

389422500 389420010
ACRO389422500EA 0 CNY
ACRO389422500 ACRO389420010
酒石酸氢钠 98+% for analysis
酒石酸氢钠
分子式: C₄H₅NaO₆
分子量: 172.07 g/mol
溶点: 253 °C
存储条件: Ambient
MDL编号: MFCD00065393
CAS号: 526-94-3

现在订购

产品技术规格

Appearance White Powder
pH 3.0 to 3.6 (5 % solution)
Titration with NaOH ≥98.0 %
Chloride (Cl) ≤0.02 %
Sulfate (SO4) ≤0.02 %
Heavy metals (as Pb) ≤0.002 %
Insoluble matter ≤0.03 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR