乙烯基氯化镁 16,5% w/w 1.9 M THF, AcroSeal®

供应商: Thermo Fisher Scientific
Danger

Synonyms: Magnesium,chloroethenyl-

252598000 252591000
ACRO252598000EA 0 CNY
ACRO252598000 ACRO252591000
乙烯基氯化镁 16,5% w/w 1.9 M THF, AcroSeal®
乙烯基氯化镁
分子式: C₂H₃ClMg
分子量: 86.801 g/mol
沸点: 66 °C
密度: 0.98 g/cm³
闪点: –17 °C
存储条件: Ambient
MDL编号: MFCD00013594
CAS号: 3536-96-7
UN号: 3399
ADR: 4.3 I

现在订购

产品技术规格

Appearance Clear to hazy yellow to dark brown Solution
Titration Active base 1.68 to 1.72 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR