Combined laboratory refrigerators/freezers, Barnstead
供应商: Thermo Fisher Scientific
3551-11 3560-1A 3566-11ACN
BARN3551-11EA 0 CNY
BARN3551-11 BARN3560-1A BARN3566-11ACN
Combined laboratory refrigerators/freezers, Barnstead
冰箱
Designed for the storage of frozen biological and laboratory samples. For laboratory and general use where there is a concern with volatile or flammable materials.
  • Spark-free interior and exteriors reduce the risk of an explosion
  • Manual defrost
  • Tough white exteriors
  • CFC- and HCFC-free