1-溴-2-丁酮 90% stabilised

供应商: Thermo Fisher Scientific
Warning

Synonyms: 溴丁酮

222450010 222450050
ACRO222450010EA 0 CNY
ACRO222450010 ACRO222450050
1-溴-2-丁酮 90% stabilised
1-溴-2-丁酮
分子式: BrH₂CC(O)CH₂CH₃
分子量: 151 g/mol
沸点: 105 °C (150 mmHg)
密度: 1.479 g/cm³
闪点: 68 °C
存储条件: Refrigerator
MDL编号: MFCD00000207
CAS号: 816-40-0
UN号: 3334
ADR: 9 III

现在订购

产品技术规格

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4650 to 1.4690 (20°C, 589 nm)
GC ≥88 %
Stabilizer Approx. 0.5 % CaCO3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR