Propyl-p-toluenesulphonate 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
p-Toluenesulphonic acid Propyl Ester
380600250
380601000
ACRO380600250EA
0
CNY
ACRO380600250
ACRO380601000
Propyl-p-toluenesulphonate 99%
Propyl-p-toluenesulphonate
Appearance: Clear light yellow Liquid
分子式:
C₁₀H₁₄O₃S 分子量: 214.28 g/mol 沸点: 140 °C (2 mmHg) 密度: 1.15 g/cm³ 存储条件: Ambient |
MDL编号:
MFCD00059244 CAS号: 599-91-7 |
产品技术规格
Appearance | Clear light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5065 to 1.5085 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...