Butyl-p-toluenesulphonate 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Butyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid Butyl Ester
380590250
380591000
ACRO380590250EA
0
CNY
ACRO380590250
ACRO380591000
Butyl-p-toluenesulphonate 99%
Butyl-p-toluenesulphonate
CAS No.: 778-28-9
分子式:
C₁₁H₁₆O₃S 分子量: 228.31 g/mol 沸点: 170…172 °C (10 mmHg) 密度: 1.12 g/cm³ 存储条件: Ambient |
MDL编号:
MFCD00027203 CAS号: 778-28-9 |
产品技术规格
Appearance | Clear light yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5030 to 1.5050 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...