氧化三乙氧基钒(V) 95%

供应商: Thermo Fisher Scientific
Warning

Synonyms: Triethoxyoxovanadium, Triethyl vanadate

380010050 380010250
ACRO380010050EA 0 CNY
ACRO380010050 ACRO380010250
氧化三乙氧基钒(V) 95%
氧化三乙氧基钒(V)

Appearance: Yellow-green to dark green Liquid

分子式: C₆H₁₈O₄V
沸点: 125…128 °C (760 mmHg)
密度: 1,139 g/cm³
闪点: 71 °C
存储条件: Ambient
MDL编号: MFCD00799170
CAS号: 1686-22-2
UN号: 3285
ADR: 6.1,III

现在订购

产品技术规格

Appearance Dark yellow to dark green to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5080 to 1.5130 (20°C, 589 nm)
Titration with KMnO4 ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR