Methyl L-tryptophanate hydrochloride 98%
供应商: Thermo Fisher Scientific
Synonyms:
H-Trp-OMe.HCl, H-Trp-OMe·HCl, L-Tryptophan methyl ester hydrochloride
376650050
376650250
ACRO376650050EA
0
CNY
ACRO376650050
ACRO376650250
Methyl L-tryptophanate hydrochloride 98%
Methyl L-tryptophanate hydrochloride
Appearance: White to off-white Powder
分子式:
C₁₂H₁₄N₂O₂·HCl 分子量: 254.7 g/mol 溶点: 218…220 °C 存储条件: Refrigerator |
MDL编号:
MFCD00066134 CAS号: 7524-52-9 |
产品技术规格
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % |
Specific optical rotation | +17° to +19° (20°C, 589 nm) (c=5, CH3OH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...