(S)-(+)-5-羟甲基-2-吡咯烷酮对甲苯磺酸酯 95%

供应商: Thermo Fisher Scientific
Warning

Synonyms: (S)-5-(Hydroxymethyl)pyrrolidin-2-one 4-methylbenzenesulphonate, (-)-L-Pyroglutamol p-toluenesulphonate, Toluene-4-sulphonic acid (S)-5-oxo-pyrrolidin-2-ylmethyl ester, (R)-(5-Oxopyrrolidin-2-yl)methyl 4-methylbenzenesulphonate

348410010 348410050
ACRO348410010EA 0 CNY
ACRO348410010 ACRO348410050
(S)-(+)-5-羟甲基-2-吡咯烷酮对甲苯磺酸酯 95%
(S)-(+)-5-羟甲基-2-吡咯烷酮对甲苯磺酸酯

CAS No.: 51693-17-5

分子式: C₁₂H₁₅NO₄S
分子量: 269.32 g/mol
溶点: 122…129 °C
存储条件: Refrigerator
MDL编号: MFCD00274229
CAS号: 51693-17-5

现在订购

产品技术规格

Appearance Colorless to beige Powder
Infrared spectrum Conforms
Melting point 122 to 129 °C
HPLC ≥94 %
Specific optical rotation +8° to +9° (25 °C, 589 nm) (c=1, C2H5OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR