(S)-(-)-2-(二苯甲基)吡咯烷 97+%

供应商: Thermo Fisher Scientific
Warning

Synonyms: (S)-2-Diphenylmethylpyrrolidine

348370010 348370050
ACRO348370010EA 0 CNY
ACRO348370010 ACRO348370050
(S)-(-)-2-(二苯甲基)吡咯烷 97+%
(S)-(-)-2-(二苯甲基)吡咯烷

CAS No.: 119237-64-8

分子式: C₁₇H₁₉N
密度: 1,062 g/cm³
闪点: 109 °C
存储条件: Refrigerator
MDL编号: MFCD00799525
CAS号: 119237-64-8

现在订购

产品技术规格

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5860 to 1.5880 (20°C, 589 nm)
GC ≥97.0 %
Specific optical rotation -2° to -4° (25°C, 589 nm) (c=1, chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR