丙戊酸 98%
供应商: Thermo Fisher Scientific
|
Danger
|
271830050
271830250
ACRO271830050EA
0
CNY
ACRO271830050
ACRO271830250
丙戊酸 98%
丙戊酸
Appearance: White Powder
分子式:
C₈H₁₅O₂Na 溶点: >300 °C 存储条件: Ambient |
MDL编号:
MFCD00078604 CAS号: 1069-66-5 |
产品技术规格
Appearance | White Powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...