Benzyl dimethylphosphonoacetate 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: Dimethylphosphonoacetic Acid Benzyl Ester, Dimethyl (Cbz-methyl)phosphonate, Benzyl 2-(dimethoxyphosphoryl)acetate, Dimethyl (Benzyloxycarbonylmethyl)phosphonate

387370050 387370250
ACRO387370050EA 0 CNY
ACRO387370050 ACRO387370250
Benzyl dimethylphosphonoacetate 98%
Benzyl dimethylphosphonoacetate

Appearance: Clear colorless to light brown Liquid

分子式: C₁₁H₁₅O₅P
分子量: 258.21 g/mol
沸点: 148…150 °C (.1 mmHg)
存储条件: Ambient
MDL编号: MFCD01872507
CAS号: 57443-18-2

现在订购

产品技术规格

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5050 to 1.5070 (20°C, 589 nm)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR