反式-苄基双(三苯基膦)氯化钯(II)
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
苄基双(三苯基膦)氯化钯(II)
369711000
369715000
ACRO369711000EA
0
CNY
ACRO369711000
ACRO369715000
反式-苄基双(三苯基膦)氯化钯(II)
反式-苄基双(三苯基膦)氯化钯(II)
Appearance: Yellow Crystalline powder
分子式:
C₄₃H₃₇ClP₂Pd 分子量: 757.59 g/mol 溶点: 166…170 °C 存储条件: Ambient |
MDL编号:
MFCD00010395 CAS号: 22784-59-4 |
产品技术规格
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Palladium (Pd) | ≥13.8 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...