二氯苯基钌(II)二聚体 97%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Dichloro(benzene)ruthenium(II) dimer
367480010
367480050
ACRO367480010EA
0
CNY
ACRO367480010
ACRO367480050
二氯苯基钌(II)二聚体 97%
二氯苯基钌(II)二聚体
CAS No.: 37366-09-9
分子式:
C₁₂H₁₂Cl₄Ru₂ 分子量: 500.17 g/mol 溶点: 249 °C 存储条件: Ambient |
MDL编号:
MFCD00064686 CAS号: 37366-09-9 |
产品技术规格
Appearance | Red to brown Powder or crystals |
Infrared spectrum | Conforms |
Ruthenium (Ru) | 39.2 to 41.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...