Fast Blue BB base 98%, pure

供应商: Thermo Fisher Scientific
Danger

Synonyms: 4'-Amino-2',5'-diethoxybenzanilide, N-(4-Amino-2,5-diethoxyphenyl)benzamide, Azoic Diazo No. 20, Fast blue BB, C.I. 37175, Azoic diazo component 20 (base)

191570250 191571000
ACRO191570250EA 0 CNY
ACRO191570250 ACRO191571000
Fast Blue BB base 98%, pure
Fast Blue BB base

Appearance: Grey to brown Powder and/or chunks

分子式: C₁₇H₂₀N₂O₃
分子量: 300.36 g/mol
溶点: 98…100 °C
存储条件: Ambient
MDL编号: MFCD00009091
CAS号: 120-00-3
UN号: 3143
ADR: 6.1 II

现在订购

产品技术规格

Appearance Grey to brown Powder and/or chunks
Infrared spectrum Conforms
UV spectrum Complies
Water ≤1 % (K.F.)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR