4-甲酰肉桂酸,主要为反式 predominantly trans 98%

供应商: Thermo Fisher Scientific
Warning

176590010 176590050
ACRO176590010EA 0 CNY
ACRO176590010 ACRO176590050
4-甲酰肉桂酸,主要为反式 predominantly trans 98%
4-甲酰肉桂酸,主要为反式

CAS No.: 23359-08-2

分子式: C₁₀H₈O₃
溶点: 250…255 °C (dec.)
存储条件: Ambient
MDL编号: MFCD00006955
CAS号: 23359-08-2

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR