5-氯-2-戊酮乙烯缩酮 97%
供应商: Thermo Fisher Scientific
Synonyms:
2-(3-氯丙基)-2-甲基-1,3-二氧戊环
166040050
166040250
ACRO166040050EA
0
CNY
ACRO166040050
ACRO166040250
5-氯-2-戊酮乙烯缩酮 97%
5-氯-2-戊酮乙烯缩酮
Appearance: Clear colorless to light yellow Liquid
分子式:
C₇H₁₃ClO₂ 分子量: 164.629 g/mol 沸点: 73…76 °C (7 mmHg) 密度: 1.09 g/cm³ 闪点: 72 °C 存储条件: Ambient |
MDL编号:
MFCD00003217 CAS号: 5978-08-5 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4490 to 1.4510 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...