5-Methyl-DL-tryptophan 98%
供应商: Thermo Fisher Scientific
Synonyms:
2-Amino-3-(5-methyl-1H-indol-3-yl)propanoic acid, H-DL-Trp(5-Me)-OH
127970010
127972500
ACRO127970010EA
0
CNY
ACRO127970010
ACRO127972500
5-Methyl-DL-tryptophan 98%
5-Methyl-DL-tryptophan
CAS No.: 951-55-3
分子式:
C₁₂H₁₄N₂O₂ 分子量: 218.26 g/mol 溶点: 280…282 °C (dec.) 存储条件: Ambient |
MDL编号:
MFCD00005652 CAS号: 951-55-3 |
产品技术规格
Appearance | Off-white Powder |
Infrared spectrum | Conforms |
Water | ≤1 % (K.F.) |
Sulfated ash | ≤0.5 % |
Appearance of solution | (5% soln. in 1 N NaOH) clear, pale yellowish |
Titration with HClO4 | ≥97.5 % |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...