甲基2-羟基异丁酸酯 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-羟基异丁酸甲酯, α-羟基异丁酸甲酯
126970050
126970500
ACRO126970050EA
0
CNY
ACRO126970050
ACRO126970500
甲基2-羟基异丁酸酯 99%
甲基2-羟基异丁酸酯
Appearance: Clear colorless Liquid
分子式:
C₅H₁₀O₃ 分子量: 118.13 g/mol 沸点: 137 °C 密度: 1.03 g/cm³ 闪点: 41 °C 存储条件: Ambient |
MDL编号:
MFCD00004457 CAS号: 2110-78-3 UN号: 3272 ADR: 3 III |
产品技术规格
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4100 to 1.4120 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...