3-(Hydroxymethyl)indole 97%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
3-Indolmethanol, 3-Indolcarbinol, (1H-Indol-3-yl)methanol, Indole-3-methanol, (Indole-3-yl)methanol, 3-Indolemethanol, Indole-3-carbinol
122190050
122190250
ACRO122190050EA
0
CNY
ACRO122190050
ACRO122190250
3-(Hydroxymethyl)indole 97%
3-(Hydroxymethyl)indole
分子式:
C₉H₉NO 分子量: 147.18 g/mol 溶点: 92…94 °C 存储条件: Refrigerator |
MDL编号:
MFCD00005632 CAS号: 700-06-1 |
产品技术规格
Appearance | Off-white to yellow to light orange Crystalline powder or flakes |
Infrared spectrum | Conforms |
Melting point | 92 to 100 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...