1,2,3,4,6-α-D(+)-葡萄糖五乙酸酯 98%
供应商: Thermo Fisher Scientific
Synonyms:
1,2,3,4,6-五-O-乙酰基-α-D-吡喃葡萄糖
119910250
119911000
ACRO119910250EA
0
CNY
ACRO119910250
ACRO119911000
1,2,3,4,6-α-D(+)-葡萄糖五乙酸酯 98%
1,2,3,4,6-α-D(+)-葡萄糖五乙酸酯
Appearance: White to off-white Powder
分子式:
C₁₆H₂₂O₁₁ 溶点: 109…113 °C 闪点: >150 °C 存储条件: Ambient |
MDL编号:
MFCD00064071 CAS号: 604-68-2 |
产品技术规格
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Specific optical rotation | +96.5° to +103.5° (20°C, 589 nm) (c=1, CHCl3) |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...