4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride 97%

供应商: Thermo Fisher Scientific
Danger

109930010 109930050 109930250
ACRO109930010EA 0 CNY
ACRO109930010 ACRO109930050 ACRO109930250
4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride 97%
4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride

Appearance: White to light yellow Crystalline powder

分子式: C₁₁H₁₃Cl₂N
分子量: 230.132 g/mol
溶点: 199…204 °C
存储条件: Ambient
MDL编号: MFCD00012753
CAS号: 51304-61-1
UN号: 2811
ADR: 6.1 II

现在订购

产品技术规格

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 199 to 204 °C
Titration with HClO4 ≥96.0 %
Distillation range 170 to 195 °C
Aromatics ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR