3,4-Epoxytetrahydrofuran 96%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 3,6-Dioxabicyclo[3.1.0]hexane

425530010 425530100
ACRO425530010EA 0 CNY
ACRO425530010 ACRO425530100
3,4-Epoxytetrahydrofuran 96%
3,4-Epoxytetrahydrofuran

CAS No.: 285-69-8

分子式: C₄H₆O₂
分子量: 86.09 g/mol
密度: 1.237 g/cm³
闪点: >38 °C
存储条件: Refrigerator
MDL编号: MFCD00800639
CAS号: 285-69-8
UN号: 1993
ADR: 3 III

现在订购

产品技术规格

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4490 (20°C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR