Sodium polyanetholesulphonate
供应商: Thermo Fisher Scientific
|
Warning
|
419630250
419631000
ACRO419630250EA
0
CNY
ACRO419630250
ACRO419631000
Sodium polyanetholesulphonate
Sodium polyanetholesulphonate
CAS No.: 55963-78-5
分子式:
(C₁₀H₁₁O₄SNa)n 存储条件: Ambient |
MDL编号:
MFCD00148427 CAS号: 55963-78-5 |
产品技术规格
Appearance | White to yellow to beige to tan Powder or crystals |
Infrared spectrum | Conforms |
pH | 5.0 to 7.0 (1 % in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...