DNA sodium salt (Deoxyribonucleic acid sodium salt), M.W. 50000-100000 Da, 纯净

供应商: Thermo Fisher Scientific

406250250 406251000
ACRO406250250EA 0 CNY
ACRO406250250 ACRO406251000
DNA sodium salt (Deoxyribonucleic acid sodium salt), M.W. 50000-100000 Da, 纯净
DNA sodium salt (Deoxyribonucleic acid sodium salt)

CAS No.: 68938-01-2


Appearance: Off-white to tan Powder

存储条件: Ambient
MDL编号: MFCD00130922
CAS号: 68938-01-2

现在订购

产品技术规格

Appearance Almost white to light yellow to tan Powder
Infrared spectrum Conforms
Loss on drying ≤18 %
Protein ≤5 %
Phosphorus (P) ≤9 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR