Brucine sulphate heptahydrate 98+% ACS
供应商: Thermo Fisher Scientific
|
Danger
|
403290100
403290250
ACRO403290100EA
0
CNY
ACRO403290100
ACRO403290250
Brucine sulphate heptahydrate 98+% ACS
Brucine sulphate heptahydrate
Appearance: White to off-white Crystalline powder
分子式:
C₂₃H₂₆N₂O₄ 存储条件: Ambient |
MDL编号:
MFCD00150159 CAS号: 60583-39-3 UN号: 2811 ADR: 6.1,I |
产品技术规格
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤13.0 % (1 g, 105°C, 6 h) |
Residue after ignition | ≤0.1 % |
Titration with HClO4 | ≥98.0 % (on dry substance) |
Sensitivity | (to nitrate) Passes test |
Clarity of solution | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...