Dimethyl(2-hydroxyethyl)phosphonate 95%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(2-Hydroxyethyl)phosphonic Acid Dimethyl Ester
338160050
338160250
ACRO338160050EA
0
CNY
ACRO338160050
ACRO338160250
Dimethyl(2-hydroxyethyl)phosphonate 95%
Dimethyl(2-hydroxyethyl)phosphonate
Appearance: Clear colorless to light yellow Liquid
分子式:
C₄H₁₁O₄P 分子量: 154.10 g/mol 沸点: 98…104 °C (.1 mmHg) 密度: 1.247 g/cm³ 闪点: 192 °C 存储条件: Ambient |
MDL编号:
MFCD00043376 CAS号: 54731-72-5 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4410 to 1.4460 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...