2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride 98%

供应商: Thermo Fisher Scientific

Synonyms: 2-((Dimethylamino)methyl)cyclohexanone hydrochloride, Tramadol Related Compound B

335620010 335620050
ACRO335620010EA 0 CNY
ACRO335620010 ACRO335620050
2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride 98%
2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride

Appearance: White Crystalline powder

分子式: C₉H₁₈ClNO
分子量: 191.699 g/mol
溶点: 147…151 °C
存储条件: Ambient
MDL编号: MFCD00102580
CAS号: 42036-65-7

现在订购

产品技术规格

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR