Ethyl 1-hydroxycyclopropanecarboxylate 90%

供应商: Thermo Fisher Scientific

Synonyms: Ethyl 1-hydroxycyclopropanecarboxylate

335300010 335300050
ACRO335300010EA 0 CNY
ACRO335300010 ACRO335300050
Ethyl 1-hydroxycyclopropanecarboxylate 90%
Ethyl-1-hydroxycyclopropanecarboxylate

CAS No.: 137682-89-4

分子式: C₆H₁₀O₃
分子量: 130.143 g/mol
沸点: 78 °C (15 mmHg)
存储条件: Refrigerator
MDL编号: MFCD01863257
CAS号: 137682-89-4

现在订购

产品技术规格

Appearance Clear light yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4470 (20°C, 589 nm)
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR