Tris(dibenzylideneacetone)dipalladium(0) 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: Pd2(dba)3

318775000 318770050
ACRO318775000EA 0 CNY
ACRO318775000 ACRO318770050
Tris(dibenzylideneacetone)dipalladium(0) 97%
三(二亚苄基丙酮)二钯(0)

CAS No.: 51364-51-3

分子式: C₅₁H₄₂O₃Pd₂
分子量: 915.72 g/mol
溶点: 152…155 °C
存储条件: Ambient
MDL编号: MFCD00013310
CAS号: 51364-51-3

现在订购

产品技术规格

Appearance Purple to black Crystalline powder
Infrared spectrum Conforms
Elemental Analysis ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR